Acetone ≥99.5%, Cleanroom® LP for the electronics industry
Supplier: KMG
|
Danger
|
Synonyms:
2-Propanone
, DMK
, Propanone
, Dimethyl ketone
200438
71009-508EA
4158.17
USD
71009-508
Acetone ≥99.5%, Cleanroom® LP for the electronics industry
Acetone
Formula:
CH₃COCH₃ MW: 58.08 g/mol Boiling Pt: 56.2 °C (1013 hPa) Melting Pt: –95.4 °C Density: 0.792 g/cm³ (20 °C) Flash Pt: <–20 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00008765 CAS Number: 67-64-1 EINECS: 200-662-2 UN: 1090 ADR: 3,II REACH: 01-2119471330-49 |
Specification Test Results
Assay | 99.5 - 100% |
Methanol | 0 - 0.05% |
Color APHA | <10 AP |
Acidity | <0.3 µeq/g |
Alkalinity | <0.3 µeq/g |
Water | 0 - 0.5% |
Aldehydes | 0 - 0.002% |
Residue After Evaporation | <5000 ppb |
Phosphate (PO₄) | <50 ppb |
Chloride (Cl) | <200 ppb |
Aluminum (Al) | <50 ppb |
Antimony (Sb) | <5 ppb |
Arsenic (As) | <5 ppb |
Barium (Ba) | <50 ppb |
Beryllium (Be) | <50 ppb |
Bismuth (Bi) | <50 ppb |
Boron (B) | <50 ppb |
Cadmium (Cd) | <50 ppb |
Calcium (Ca) | <50 ppb |
Chromium (Cr) | <50 ppb |
Cobalt (Co) | <10 ppb |
Copper (Cu) | <10 ppb |
Gallium (Ga) | <50 ppb |
Germanium (Ge) | <50 ppb |
Gold (Au) | <50 ppb |
Iron (Fe) | <50 ppb |
Lead (Pb) | <50 ppb |
Lithium (Li) | <50 ppb |
Magnesium (Mg) | <50 ppb |
Manganese (Mn) | <50 ppb |
Molybdenum (Mo) | <50 ppb |
Nickel (Ni) | <10 ppb |
Niobium (Nb) | <50 ppb |
Potassium (K) | <50 ppb |
Silicon (Si) | <50 ppb |
Silver (Ag) | <50 ppb |
Sodium (Na) | <50 ppb |
Strontium (Sr) | <50 ppb |
Tantalum (Ta) | <50 ppb |
Thallium (Tl) | <50 ppb |
Tin (Sn) | <50 ppb |
Titanium (Ti) | <50 ppb |
Vanadium (V) | <50 ppb |
Zinc (Zn) | <50 ppb |
Zirconium (Zr) | <50 ppb |
0.2 Micron Particle Count | <200 Par/ml |
0.3 Micron Particle Coun | <200 Par/ml |
0.5 Micron Particle Count | <150 Par/ml |
1.0 Micron Particle Count | <25 Par/ml |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...