Giemsa's stain, powder, HARLECO®, certified by the Biological Stain Commission for hematology, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
Synonyms:
Giemsa's azur-eosin-methylene blue
, Azure mixture sicc. Giemsa stain
, Azur-Eosin-Methylene blue acc. Giemsa
228-12
228-12
15204-122EA
527.05
USD
CA15204-122
15204-122
Giemsa's stain, powder, HARLECO®, certified by the Biological Stain Commission for hematology, Sigma-Aldrich®
Giemsa's stain
Storage Temperature:
Ambient |
MDL Number:
MFCD00081642 CAS Number: 51811-82-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...