Brilliant cresyl blue solution 1% in saline

Supplier: AZER SCIENTIFIC INC

Synonyms: BCBBrilliant Blue CBrilliant cresyl blue zinc chloride double salt1,3-Diamino-7-(diethylamino)-3-methylphenoxazin-5-ium tetrachlorozincate (2:1)CI 51010

ES8002-125
89130-182EA 45.14 USD
89130-182
Brilliant cresyl blue solution 1% in saline
Brilliant Cresyl Blue

For the staining of reticulocytes and tricomonads.


  • For use on formalin-fixed, paraffin-embedded, or frozen sections
  • Used on blood smears and bone marrow


These components are intended for use in the visualization of cells present in hematopoietic tissues and certain microorganisms.

Formula: C₃₄H₄₂Cl₄N₈O₂Zn
MW: 801.96 g/mol
MDL Number: MFCD00148901
CAS Number: 81029-05-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR