Basic fuchsin working solution for Gram staining

Supplier: Electron Microscopy Sciences
Warning

Synonyms: Basic-fuchsinDiamondfuchsinBasic Red 9(4-(4-Aminophenyl)(4-iminocyclohexa-2,5-dienylidene)methyl)-2-methylaniline hydrochlorideBasic fuchsinFuchsine base monohydrochlorideRosaniline hydrochloridePararosanilineFuchsin basicBasic fuchsin hydrochlorideCI 425104-((4-Aminophenyl)(4-iminocyclohexa-2,5-dien-1-ylidene)methyl)-2-methylaniline hydrochlorideMagenta O

26105-3A
101411-042EA 25.4 USD
101411-042
Basic fuchsin working solution for Gram staining
Basic Fuchsin

Brown and Brenn Method reagent for Gram Positive and Gram Negative Bacteria


Gram Negative Bacteria for bacteria stain: Gram + / Gram – bacteria and filaments of nocardia, etc


Stain results - Gram + Bacteria, Nocardia and Actinomyces Filaments: Blue, Gram – Bacteria, Nuclei: Red, Additional tissue elements: Yellow

Formula: C₂₀H₂₀ClN₃
MW: 337.85 g/mol
MDL Number: MFCD00012569
CAS Number: 632-99-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR