5(6)-Carboxyfluorescein N-hydroxysuccinimide ester (5/6-FAM SE)
Supplier: CHEM IMPEX INTL INC MS
Synonyms:
5(6)-FAM SE
, NHS-Fluorescein
, FLUOS
22981-25MG
22981-100MG
102624-514EA
255.28
USD
102624-514
102568-818
5(6)-Carboxyfluorescein N-hydroxysuccinimide ester (5/6-FAM SE)
5(6)-Carboxyfluorescein N-hydroxysuccinimide ester (5/6-FAM SE)
Formula:
C₂₅H₁₅NO₉ MW: 473.39 g/mol |
MDL Number:
MFCD04119713 CAS Number: 117548-22-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...