α-Naphtholphthalein indicator, Fluka™

Supplier: Honeywell Research Chemicals
Warning

Synonyms: 3,3-Bis(4-hydroxynaphthalen-1-yl)isobenzofuran-1(3H)-one

33424-10G
BJ33424-10GEA 627.73 USD
BJ33424-10G
α-Naphtholphthalein indicator, Fluka™
α-Naphtholphthalein
Formula: C₂₈H₁₈O₄
MW: 418.45 g/mol
Melting Pt: 238…240 °C
Storage Temperature: Ambient
MDL Number: MFCD00036202
CAS Number: 596-01-0
EINECS: 209-875-5

Order Now

Specification Test Results

Change range (pH 7.1-8.3) complying
Test of sensitivity complying

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR