Xylenol orange tetrasodium salt Indicator_Metal_titration, Fluka™

Supplier: Honeywell Research Chemicals

Synonyms: Xylenol orange water soluble, 2,2',2'',2'''-((((1,1-Dioxido-3H-benzo[c][1,2]oxathiole-3,3-diyl)bis(6-hydroxy-5-methyl-3,1-phenylene))bis(methylene))bis(azanetriyl))tetraacetic acid, sodium salt (1:4)

33825-5G 33825-1G
BJ33825-5GEA 217.39 USD
BJ33825-5G BJ33825-1G
Xylenol orange tetrasodium salt Indicator_Metal_titration, Fluka™
Xylenol orange tetrasodium salt

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Formula: C₃₁H₂₈N₂Na₄O₁₃S
MW: 760.59 g/mol
Melting Pt: 210
Storage Temperature: Ambient
MDL Number: MFCD00044293
CAS Number: 3618-43-7
EINECS: 222-805-8

Order Now

Specification Test Results

Identity (IR) complying
Change range complying
Sulfated ash Max. 50 %
Absorption maximum 575 - 585 nm
Absorbtion (1%/1cm) Min. 400
Suitability for Complexometry complying
Test of sensitivity complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR