β-tri-Calcium phosphate (β-phase) ≥98% (β-phase basis), sintered powder, Puriss. p.a., Fluka™

Supplier: Honeywell Research Chemicals

49963-100G 49963-10G
BJ49963-100GEA 1007.65 USD
BJ49963-100G BJ49963-10G
β-tri-Calcium phosphate (β-phase) ≥98% (β-phase basis), sintered powder, Puriss. p.a., Fluka™
β-tri-Calcium phosphate (β-phase)
Formula: Ca₃O₈P₂
MW: 310.18 g/mol
Melting Pt: 1.670 °C
Density: 3.27 g/cm³ (20.5 °C)
Storage Temperature: Ambient
MDL Number: MFCD00015984
CAS Number: 7758-87-4
EINECS: 231-840-8
Merck Index: 12,01741

Order Now

Specification Test Results

Phase-composition (XRD) complying
Arsenic (As) Max. 0.5 ppm
Barium (Ba) Max. 20 ppm
Cadmium (Cd) Max. 1 ppm
Cobalt (Co) Max. 1 ppm
Chromium (Cr) Max. 10 ppm
Copper (Cu) Max. 5 ppm
Mercury (Hg) Max. 500 ppb
Nickel (Ni) Max. 5 ppm
Lead (Pb) Max. 5 ppm
Zinc (Zn) Max. 20 ppm
Total S (as SO₄) Max. 500 ppm
Heavy metals (as Pb) Max. 50 ppm
Chloride (Cl) Max. 100 ppm

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR