Sulfuric acid 95.0-98.0% ACS, Fluka™

Supplier: Honeywell Research Chemicals
Danger

320501-1L
BJ320501-1LEA 138.18 USD
BJ320501-1L
Sulfuric acid 95.0-98.0% ACS, Fluka™
Sulfuric acid
Formula: H₂SO₄
MW: 98.08 g/mol
Boiling Pt: 310 °C (1.013 hPa)
Melting Pt: -14 to -10 °C
Density: 1.840 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 1830
ADR: 8,II

Order Now

Specification Test Results

Assay 95.0 - 98.0 %
Density (D 20/4) 1.825 - 1.836
Residue on ignition Max. 5 ppm
Ammonium (NH₄) Max. 2 ppm
Arsenic (As) Max. 0.01 ppm
Iron (Fe) Max. 0.2 ppm
Mercury (Hg) Max. 0.005 ppm
Heavy metals (as Pb) Max. 1 ppm
Chloride (Cl) Max. 0.2 ppm
Nitrate (NO₃) Max. 0.5 ppm
KMnO₄ reducing matter (as SO₂) Max. 2 ppm
APHA Max. 10
Appearance complying
Appearance Free from suspended or insoluble matter

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR