Sulfuric acid 95-97, Puriss., meetSpec_BP,Ph.Eur., Fluka™
Supplier: Honeywell Research Chemicals
|
Danger
|
07208-6X1L
07208-5L
07208-4X5L
07208-4X2.5L
BJ07208-1LCS
568.9
USD
BJ07208-1L
BJ07208-2.5L
BJ07208-5L
BJ072082.5LGL
BJ07208-4X5L
BJ07208-4X2.5L
Sulfuric acid 95-97, Puriss., meetSpec_BP,Ph.Eur., Fluka™
Sulfuric acid
Formula:
H₂SO₄ MW: 98.08 Boiling Pt: 330 Melting Pt: 10.38 Density: 1.840 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00064589 CAS Number: 7664-93-9 EINECS: 231-639-5 UN: 1830 ADR: 8 II |
Specification Test Results
Assay | 95.0 - 97.0 % |
Residue on ignition | Max. 0.0005 % |
Arsenic (As) | Max. 0.1 ppm |
Calcium (Ca) | Max. 0.001 % |
Cadmium (Cd) | Max. 0.0005 % |
Cobalt (Co) | Max. 0.0005 % |
Chromium (Cr) | Max. 0.0005 % |
Copper (Cu) | Max. 0.0005 % |
Iron (Fe) | Max. 2 ppm |
Potassium (K) | Max. 0.005 % |
Magnesium (Mg) | Max. 0.0005 % |
Manganese (Mn) | Max. 0.0005 % |
Sodium (Na) | Max. 0.005 % |
Nickel (Ni) | Max. 0.0005 % |
Lead (Pb) | Max. 0.0005 % |
Zinc (Zn) | Max. 0.0005 % |
Heavy metals (as Pb) | Max. 5 ppm |
Chloride (Cl) | Max. 1 ppm |
Nitrate (NO₃) | complying |
Appearance of the solution | complying |
Residual solvents | complying |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...