Sodium sulfate, anhydrous ≥99.0%, Puriss. p.a. Reag. Ph. Eur., ACS, ISO, Fluka™

Supplier: Honeywell Research Chemicals

Synonyms: Disodium sulfate, Bisodium sulfate, Disodium sulphate, Sulfuric acid disodium salt, Bisodium sulphate, Sulphuric acid disodium salt

31481-6X1KG 31481-6X2.5KG 31481-500G
BJ31481-1KGCS 696.68 USD
BJ31481-1KG BJ31481-2.5KG BJ31481-500G
Sodium sulfate, anhydrous ≥99.0%, Puriss. p.a. Reag. Ph. Eur., ACS, ISO, Fluka™
Sodium sulphate
Formula: Na₂SO₄
Melting Pt: 884 °C
Density: 2.68 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003504
CAS Number: 7757-82-6
EINECS: 231-820-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a4977cb8ca54638f22a60675bfbef71bbb77ab6&vid=942501aaa5afc8d85b82628854aac94d517322eb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 99.0 %
Assay (calculated to the dried substance) 99 - 101.0 %
Loss on ignition (800°C) Max. 0.5 %
Loss on drying (130°C) Max. 0.5 %
Water insoluble matter Max. 0.01 %
pH (5 %, 25 °C) 5.2 - 8.0
Free acid (as H₂SO₄) Max. 0.05 %
Free alkali (as NaOH) Max. 0.03 %
Arsenic (As) Max. 0.0001 %
Calcium (Ca) Max. 20 ppm
Iron (Fe) Max. 5 ppm
Potassium (K) Max. 0.01 %
Magnesium (Mg) Max. 25 ppm
Zinc (Zn) Max. 0.02 %
Heavy metals (as Pb) Max. 5 ppm
Chloride (Cl) Max. 10 ppm
Phosphate (PO₄) Max. 0.001 %
Total N Max. 5 ppm
Appearance of the solution complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR