Sodium periodate ≥99.8%, Puriss. p.a. ACS, Reag. Ph. Eur., Fluka™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Sodium metaperiodate

30323-6X500G 30323-100G
BJ30323-500GCS 4761.79 USD
BJ30323-500G BJ30323-100G
Sodium periodate ≥99.8%, Puriss. p.a. ACS, Reag. Ph. Eur., Fluka™
Sodium periodate
Formula: NaIO₄
MW: 213.89 g/mol
Melting Pt: 270 °C (dec.)
Density: 3.86 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003534
CAS Number: 7790-28-5
EINECS: 232-197-6
UN: 3085
ADR: 5.1,I

Order Now

Specification Test Results

Assay Min. 99.8 %
Assay (excluding dried substances) 99.8 - 100.3 %
Loss on drying (110°C) Max. 0.2 %
Iron (Fe) Max. 0.001 %
Manganese (Mn) Max. 0.0001 %
Heavy metals (as Pb) Max. 0.001 %
Other halogens (as Cl) Max. 0.01 %
Sulfate (SO₄) Max. 0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR