Sodium nitrite ≥99%, Puriss. p.a. ACS, Reag. Ph. Eur., Fluka™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Diazotizing salts

31443-500G 31443-1KG 31443-250G 31443-100G
BJ31443-250GEA 87.2 USD
BJ31443-250G BJ31443-100G BJ31443-500G BJ31443-1KG
Sodium nitrite ≥99%, Puriss. p.a. ACS, Reag. Ph. Eur., Fluka™
Sodium nitrite
Formula: NaNO₂
MW: 69 g/mol
Melting Pt: 280 °C
Density: 2.17 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011118
CAS Number: 7632-00-0
EINECS: 231-555-9
UN: 1500
ADR: 5.1,III

Order Now

Specification Test Results

Assay Min. 99.0 %
Water insoluble matter Max. 0.005 %
Calcium (Ca) Max. 0.002 %
Iron (Fe) Max. 0.001 %
Potassium (K) Max. 0.005 %
Heavy metals (as Pb) Max. 0.001 %
Chloride (Cl) Max. 0.005 %
Sulfate (SO₄) Max. 0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR