Sodium metabisulfite, dry 98-100.5%, Puriss. p.a. ACS, Reag. Ph. Eur., Fluka™

Supplier: Honeywell Research Chemicals
Danger

31448-6X2.5KG 31448-500G 31448-50KG 31448-5KG
BJ31448-1KGEA 132.94 USD
BJ31448-1KG BJ31448-2.5KG BJ31448-500G BJ31448-50KG BJ31448-5KG
Sodium metabisulfite, dry 98-100.5%, Puriss. p.a. ACS, Reag. Ph. Eur., Fluka™
Sodium metabisulphite
Formula: Na₂S₂O₅
Boiling Pt: Dec. (on heating)
Melting Pt: >150 °C (dec.)
Density: 2.36 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00167602
CAS Number: 7681-57-4
EINECS: 231-673-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=110c9b08e7e977eb8c8a9a97bee16470ca9699c5&vid=004a574d6ed7e27d7cc82336f2179f23453618d0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 98.0 - 100.5 %
pH (5 %, 20°C) 3.5 - 5.0
Arsenic (As) Max. 0.5 ppm
Calcium (Ca) Max. 0.005 %
Cadmium (Cd) Max. 0.005 %
Cobalt (Co) Max. 0.005 %
Copper (Cu) Max. 0.0005 %
Iron (Fe) Max. 5 ppm
Potassium (K) Max. 0.01 %
Nickel (Ni) Max. 0.005 %
Lead (Pb) Max. 0.0005 %
Zinc (Zn) Max. 0.001 %
Heavy metals (as Pb) Max. 10 ppm
Chloride (Cl) Max. 0.005 %
Thiosulfate (S₂O₃) Max. 0.01 %
Water insoluble matter Max. 0.005 %
Appearance of the solution complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR