Sodium iodide 99-100.5% (dried basis), Puriss., meets analytical specification of BP, Ph. Eur., USP, Fluka™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: IodurilHydriodic acid sodium saltSodium monoiodide

03129-6X1KG
BJ03129-1KGCS 2215.41 USD
BJ03129-1KG
Sodium iodide 99-100.5% (dried basis), Puriss., meets analytical specification of BP, Ph. Eur., USP, Fluka™
Sodium iodide
Formula: NaI
MW: 149.89 g/mol
Boiling Pt: 1.304 °C (1.013 hPa)
Melting Pt: 662 °C
Density: 3.67 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003532
CAS Number: 7681-82-5
EINECS: 231-679-3

Order Now

Specification Test Results

Assay (calculated to the dried substance) 99.0 - 100.5 %
Loss on drying (105°C) Max. 3.0 %
Water (Karl Fischer) Max. 2.0 %
Alkalinely reacting impurities complying
Iron (Fe) Max. 5 ppm
Potassium (K) complying
Heavy metals (as Pb) Max. 5 ppm
Iodate (IO₃) Max. 2 ppm
Iodate (JO₃) complying
Sulfate (SO₄) Max. 150 ppm
Thiosulfate (S₂O₃) Max. 0.005 %
Thiosulfate and Barium complying
Nitrate, nitrite, ammonium complying
Residual solvents complying
Appearance of the solution complying

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR