Sodium hydroxide 98-100.5%, pellets, Puriss., meets analytical specification of BP, NF, Ph. Eur., E524, Fluka™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

06203-1KG 06203-5KG
BJ06203-1KGEA 135.36 USD
BJ06203-1KG BJ06203-5KG
Sodium hydroxide 98-100.5%, pellets, Puriss., meets analytical specification of BP, NF, Ph. Eur., E524, Fluka™
Sodium hydroxide
Formula: NaOH
Boiling Pt: 1.390 °C (1.013 hPa)
Melting Pt: 319 °C
Density: 2.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1823
ADR: 8,II

Order Now

Specification Test Results

Assay 98.0 - 100.5 %
Assay of Na₂CO₃ Max. 0.5 %
Aluminium (Al) Max. 0.001 %
Arsenic (As) Max. 2 ppm
Calcium (Ca) Max. 0.001 %
Iron (Fe) Max. 10 ppm
Mercury (Hg) Max. 1 ppm
Potassium (K) Max. 0.1 %
Lead (Pb) Max. 0.5 ppm
Heavy metals (as Pb) Max. 20 ppm
Chloride (Cl) Max. 100 ppm
Total N Max. 0.0005 %
Phosphate (PO₄) Max. 0.001 %
Silicate (as SiO₂) Max. 0.01 %
Sulfate (SO₄) Max. 100 ppm
Water insoluble matter + org. comp. complying
Residual solvents complying
Appearance of the solution complying
Vapor Pressure <18 mmHg ( 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR