Sodium hydrogen carbonate 99,0-100,5, powder, Puriss., meetSpec_BP,Ph.Eur.,USP,FCC,E500, Fluka™

Supplier: Honeywell Research Chemicals

Synonyms: Sodium bicarbonate

13433-6X1KG 13433-4X5KG
BJ13433-1KGCS 598.72 USD
BJ13433-1KG BJ13433-5KG
Sodium hydrogen carbonate 99,0-100,5, powder, Puriss., meetSpec_BP,Ph.Eur.,USP,FCC,E500, Fluka™
Sodium hydrogen carbonate
Formula: NaHCO₃
MW: 84.01 g/mol
Melting Pt: 270 Dec.
Density: 2.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003528
CAS Number: 144-55-8
EINECS: 205-633-8
Merck Index: 13,08655
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6b1b80d2afb3deab0c4ca836a921fcaad637890&vid=1163b37d9cf1c10fe0ed252e02a51fe12d6b50dd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.0 - 100.5 %
Assay (calculated to the dried substance) 99.0 - 100.5 %
Water insoluble matter complying
pH (5 %, 20°C) Max. 8.6
Loss on drying (via silica gel) Max. 0.2 %
Aluminium (Al) Max. 0.5 ppm
Ammonium (NH₄) Max. 20 ppm
Ammonia (NH₃) complying
Arsenic (As) Max. 2 ppm
Calcium (Ca) Max. 100 ppm
Iron (Fe) Max. 10 ppm
Mercury (Hg) Max. 1 ppm
Potassium (K) Max. 0.01 %
Lead (Pb) Max. 2 ppm
Heavy metals (as Pb) Max. 5 ppm
Carbonate (CO₃) complying
Chloride (Cl) Max. 100 ppm
Sulfate (SO₄) Max. 100 ppm
S-compounds (as SO₄) Max. 150 ppm
Appearance of the solution complying
Residual solvents complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR