Sodium chloride ≥99%, Reagent Grade

Supplier: Honeywell Research Chemicals

Synonyms: Saline, NaCl, Sea salt, Halite

S9625-10KG S9625-1KG S9625-5KG
BJS9625-10KGEA 293 USD
BJS9625-10KG BJS9625-1KG BJS9625-5KG
Sodium chloride ≥99%, Reagent Grade
Sodium chloride
Formula: NaCl
MW: 58.44 g/mol
Boiling Pt: 1413
Melting Pt: 801
Density: 2.160 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,08599

Order Now

Specification Test Results

Appearance complying
Assay (argentometric) 99.5 - 100.5 %
Colour of the solution complying
Turbidity of solution complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR