Sodium carbonate monohydrate ≥99.5% ACS, Fluka™
Supplier: Honeywell Research Chemicals
|
Warning
|
Synonyms:
Sodium carbonate hydrous
, Soda monohydrate
230952-500G
230952-2.5KG
BJ230952-2.5KGEA
276.39
USD
BJ230952-2.5KG
BJ230952-500G
Sodium carbonate monohydrate ≥99.5% ACS, Fluka™
Sodium carbonate monohydrate
Formula:
Na₂CO₃·1H₂O MW: 124.00 g/mol Melting Pt: 851 °C Density: 2.25 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149177 CAS Number: 5968-11-6 |
Specification Test Results
Assay | Min. 99.5 % |
Loss on drying (150°C) | 13.0 - 15.0 % |
Water insoluble matter | Max. 0.01 % |
Calcium (Ca) | Max. 0.03 % |
Iron (Fe) | Max. 5 ppm |
Potassium (K) | Max. 0.005 % |
Magnesium (Mg) | Max. 0.005 % |
Heavy metals (as Pb) | Max. 5 ppm |
Chloride (Cl) | Max. 0.001 % |
Phosphate (PO₄) | Max. 5 ppm |
S-compounds (as SO₄) | Max. 0.004 % |
Silicate (as SiO₂) | Max. 0.005 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...