Sodium carbonate decahydrate 99.5-100.5% (dried basis), Puriss., meets analytical specification of BP, FCC, Ph. Eur., E500, Fluka™
Supplier: Honeywell Research Chemicals
|
Warning
|
13414-6X1KG
13414-2.5KG
BJ13414-1KGCS
350.65
USD
BJ13414-1KG
BJ13414-2.5KG
Sodium carbonate decahydrate 99.5-100.5% (dried basis), Puriss., meets analytical specification of BP, FCC, Ph. Eur., E500, Fluka™
Sodium carbonate decahydrate
Formula:
Na₂CO₃·10H₂O Melting Pt: 32 °C Density: 1.460 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149178 CAS Number: 6132-02-1 EINECS: 207-838-8 |
Specification Test Results
Assay (excluding dried substance) | 99.5 - 100.5 % |
Assay of Na₂CO₃ | 36.7 - 40.0 % |
Loss on drying | 55.0 - 65.0 % |
Arsenic (As) | Max. 2 ppm |
Copper (Cu) | Max. 0.001 % |
Iron (Fe) | Max. 5 ppm |
Mercury (Hg) | Max. 1 ppm |
Potassium (K) | Max. 0.02 % |
Lead (Pb) | Max. 2 ppm |
Zinc (Zn) | Max. 0.001 % |
Heavy metals (as Pb) | Max. 10 ppm |
Chloride (Cl) | Max. 30 ppm |
Sulfate (SO₄) | Max. 100 ppm |
Appearance of the solution | complying |
Alkali hydroxide, -bicarbonates | complying |
Residual solvents | complying |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...