Sodium carbonate ≥99.5%, powder ACS, Fluka™

Supplier: Honeywell Research Chemicals
Warning

Synonyms: Soda, Carbonic acid sodium salt, Calcined soda, Disodium carbonate, Carbonic acid disodium salt, Na-X, Bisodium carbonate, Carbonic acid, disodium salt, Soda ash

223530-2.5KG 223530-500G
BJ223530-12KGEA 637.87 USD
BJ223530-12KG BJ223530-2.5KG BJ223530-500G
Sodium carbonate ≥99.5%, powder ACS, Fluka™
Sodium carbonate
Formula: Na₂CO₃
Melting Pt: 854 °C
Density: 2.53 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003494
CAS Number: 497-19-8
EINECS: 207-838-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a6ff30f1487cdef86add98b38bfa8eb3dac14f6&vid=c5b129b7acb5c897b9d970ea3130d61fb1934b54&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity (IR) complying
Assay (excluding ignited substance) Min. 99.5 %
Loss on ignition (285°C) Max. 1.0 %
Water insoluble matter Max. 0.01 %
Calcium (Ca) Max. 0.03 %
Iron (Fe) Max. 5 ppm
Potassium (K) Max. 0.005 %
Magnesium (Mg) Max. 0.005 %
Heavy metals (ICP) ACS Max. 5 ppm
Silicate (as SiO₂) Max. 0.005 %
Chloride (Cl) Max. 0.001 %
Phosphate (PO₄) Max. 0.001 %
Total S (as SO₄) Max. 0.003 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR