Sodium bromide ≥99.0% ACS, Fluka™
Supplier: Honeywell Research Chemicals
310506-500G
BJ310506-500GEA
134.34
USD
BJ310506-500G
Sodium bromide ≥99.0% ACS, Fluka™
Sodium bromide
Formula:
NaBr MW: 102.89 g/mol Boiling Pt: 1.393 °C (1.013 hPa) Melting Pt: 750 °C Density: 3.2 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003475 CAS Number: 7647-15-6 EINECS: 231-599-9 |
Specification Test Results
Assay | Min. 99.0 % |
pH (5 %, 25 °C) | 5.0 - 8.8 |
Water insoluble matter | Max. 0.005 % |
Barium (Ba) | Max. 0.002 % |
Calcium (Ca) | Max. 0.002 % |
Iron (Fe) | Max. 5 ppm |
Potassium (K) | Max. 0.1 % |
Magnesium (Mg) | Max. 0.001 % |
Heavy metals (as Pb) | Max. 5 ppm |
Bromate (BrO₃) | Max. 0.001 % |
Chloride (Cl) | Max. 0.2 % |
Sulfate (SO₄) | Max. 0.002 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...