Silver sulfate ≥99%, Puriss., Fluka™
Supplier: Honeywell Research Chemicals
|
Danger
|
Synonyms:
Disilver sulphate
, Silver(I) sulfate
, Disilver sulfate
10229-500G
BJ10229-500GEA
3432.57
USD
BJ10229-500G
Silver sulfate ≥99%, Puriss., Fluka™
Silver sulphate
Formula:
Ag₂SO₄ MW: 311.8 g/mol Boiling Pt: Dec. Melting Pt: 655 °C Density: 5.45 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003407 CAS Number: 10294-26-5 EINECS: 233-653-7 |
Specification Test Results
Assay | Min. 99 % |
Copper (Cu) | Max. 0.005 % |
Iron (Fe) | Max. 0.005 % |
Lead (Pb) | Max. 0.005 % |
Chloride (Cl) | Max. 0.001 % |
Nitrate (NO₃) | Max. 0.01 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...