Potassium peroxodisulfate ≥99.0% ACS, Fluka™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Dipotassium peroxodisulphateDipotassium peroxodisulfatePeroxydisulphuric acid dipotassium saltPotassium persulfatedi-Potassium peroxodisulfatePeroxydisulfuric acid dipotassium saltPotassium persulphatedi-Potassium peroxodisulphate

216224-500G 216224-100G
BJ216224-500GEA 116.2 USD
BJ216224-500G BJ216224-100G
Potassium peroxodisulfate ≥99.0% ACS, Fluka™
Potassium peroxodisulphate
Formula: K₂S₂O₈
MW: 270.32 g/mol
Melting Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00011386
CAS Number: 7727-21-1
EINECS: 231-781-8
UN: 1492
ADR: 5.1,III

Order Now

Specification Test Results

Assay 99.0 - 101.0 %
Water insoluble matter Max. 0.005 %
Iron (Fe) Max. 5 ppm
Manganese (Mn) Max. 1 ppm
Heavy metals (as Pb) Max. 0.001 %
Chlorine compounds (Cl) Max. 10 ppm
Vapor Density 9.3 (vs air)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR