Potassium hydroxide 85, pellets ACS, Fluka™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Potash caustic, Potassium hydrate, Caustic potash

221473-1KG 221473-2.5KG 221473-6X500G
BJ221473-12KGEA 723.45 USD
BJ221473-12KG BJ221473-1KG BJ221473-2.5KG BJ221473-25G BJ221473-3.5KG BJ221473-500G
Potassium hydroxide 85, pellets ACS, Fluka™
Potassium hydroxide
Formula: KOH
MW: 56.11 g/mol
Boiling Pt: 1320
Melting Pt: 360
Density: 2.04 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 1813
ADR: 8 II
Merck Index: 12,07806

Order Now

Specification Test Results

Assay Min. 85.0 %
Assay of K₂CO₃ Max. 2.0 %
Calcium (Ca) Max. 0.005 %
Iron (Fe) Max. 0.001 %
Magnesium (Mg) Max. 0.002 %
Sodium (Na) Max. 0.05 %
Nickel (Ni) Max. 0.001 %
Form pellets
Heavy metals (ICP) ACS Max. 0.001 %
Chloride (Cl) Max. 0.01 %
Phosphate (PO₄) Max. 5 ppm
Sulfate (SO₄) Max. 0.003 %
Total N Max. 0.001 %
Appearance complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR