Potassium hydroxide 85-100.5%, pellets, Puriss., meets analytical specification of BP, Ph. Eur., NF, Fluka™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Potash caustic, Potassium hydrate, Caustic potash

06005-6X1KG 06005-5KG 06005-4X5KG
BJ06005-1KGCS 592.85 USD
BJ06005-1KG BJ06005-5KG BJ06005-4X5KG
Potassium hydroxide 85-100.5%, pellets, Puriss., meets analytical specification of BP, Ph. Eur., NF, Fluka™
Potassium hydroxide
Formula: KOH
MW: 56.11 g/mol
Boiling Pt: 1320
Melting Pt: 360
Density: 2.04 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 1813
ADR: 8 II
Merck Index: 12,07806

Order Now

Specification Test Results

Assay 85.0 - 100.5 %
Total alkali (as KOH) Min. 85.0 %
Assay of K₂CO₃ Max. 1.0 %
Assay of Potassium (K) Min. 59.9 %
Aluminium (Al) Max. 10 ppm
Arsenic (As) Max. 0.0003 %
Calcium (Ca) Max. 0.002 %
Iron (Fe) Max. 10 ppm
Sodium (Na) Max. 1.0 %
Heavy metals (as Pb) Max. 5 ppm
Chloride (Cl) Max. 100 ppm
Phosphate (PO₄) Max. 10 ppm
Silicate (as SiO₂) Max. 0.01 %
Sulfate (SO₄) Max. 100 ppm
Total N Max. 0.0005 %
Appearance of the solution complying
Water insoluble substances complying
Residual solvents complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR