Potassium hexacyanoferrate(III) ≥99%, Puriss. p.a. Reag. Ph. Eur., ACS, ISO, Fluka™

Supplier: Honeywell Research Chemicals
Warning

Synonyms: Potassium ferricyanide(III), Tripotassium hexacyanoferrate, Potassium ferricyanide

31253-6X1KG 31253-250G 31253-500G
BJ31253-1KGCS 2328.93 USD
BJ31253-1KG BJ31253-250G BJ31253-500G
Potassium hexacyanoferrate(III) ≥99%, Puriss. p.a. Reag. Ph. Eur., ACS, ISO, Fluka™
Potassium hexacyanoferrate (III)
Formula: K₃Fe(CN)₆
MW: 329.25 g/mol
Boiling Pt: Dec. (on heating)
Melting Pt: Dec.
Density: 1.85 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011392
CAS Number: 13746-66-2
EINECS: 237-323-3
Merck Index: 13,07712

Order Now

Specification Test Results

Assay Min. 99 %
Water insoluble matter Max. 0.005 %
Calcium (Ca) Max. 0.0005 %
Cadmium (Cd) Max. 0.0005 %
Cobalt (Co) Max. 0.005 %
Copper (Cu) Max. 0.001 %
Sodium (Na) Max. 0.02 %
Nickel (Ni) Max. 0.001 %
Lead (Pb) Max. 0.002 %
Zinc (Zn) Max. 0.0005 %
Chloride (Cl) Max. 0.005 %
Hexacyanoferrate(II) ([Fe(CN)₆]₄⁻) Max. 0.02 %
Sulfate (SO₄) Max. 0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR