Potassium cyanide ≥97.0%, Puriss. p.a. Reag. Ph. Eur., ACS, Fluka™

Supplier: Honeywell Research Chemicals
Danger

31252-100G 31252-1KG 31252-250G 31252-25KG
BJ31252-100GEA 109.05 USD
BJ31252-100G BJ31252-1KG BJ31252-250G BJ31252-25KG
Potassium cyanide ≥97.0%, Puriss. p.a. Reag. Ph. Eur., ACS, Fluka™
Potassium cyanide
Formula: KCN
Melting Pt: 634.5 °C
Density: 1.56 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011397
CAS Number: 151-50-8
EINECS: 205-792-3
UN: 1680
ADR: 6.1,I

Order Now

Specification Test Results

Assay Min. 97 %
Iron (Fe) Max. 0.01 %
Sodium (Na) Max. 0.5 %
Lead (Pb) Max. 0.0002 %
Chloride (Cl) Max. 0.02 %
Phosphate (PO₄) Max. 0.005 %
Sulfate (SO₄) Max. 0.04 %
Sulfide (S) Max. 0.0005 %
Thiocyanate (SCN) Max. 0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR