Potassium carbonate, anhydrous ≥99.0% (by titrimetric analysis), Purum p.a.

Supplier: Honeywell Research Chemicals
Warning

Synonyms: Carbonic acid potassium saltPotashCarbonic acid, dipotassium salt

60110-5KG 60110-1KG
BJ60110-5KGEA 358.13 USD
BJ60110-5KG BJ60110-1KG
Potassium carbonate, anhydrous ≥99.0% (by titrimetric analysis), Purum p.a.
Potassium carbonate

  • Products with a guarantee certificate and/or suitable for the stated analytical application.

Formula: K₂CO₃
MW: 138.21 g/mol
Boiling Pt: Decomposition
Melting Pt: 891 °C
Density: 2.430 g/cm³ (19 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011382
CAS Number: 584-08-7
EINECS: 209-529-3
https://us.vwr.com/stibo/web/std.lang.all/05/98/16640598.jpg

Order Now

Specification Test Results

Assay 99.0 - 101.0 %
Calcium (Ca) Max. 50 ppm
Cadmium (Cd) Max. 50 ppm
Cobalt (Co) Max. 50 ppm
Copper (Cu) Max. 50 ppm
Iron (Fe) Max. 50 ppm
Sodium (Na) Max. 5000 ppm
Nickel (Ni) Max. 50 ppm
Lead (Pb) Max. 50 ppm
Zinc (Zn) Max. 50 ppm
Total S (as SO₄) Max. 50 ppm
Chloride (Cl) Max. 50 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR