Potassium bromide ≥99% (trace metals basis), FT-IR Grade, Fluka™
Supplier: Honeywell Research Chemicals
221864-100G
221864-25G
BJ221864-100GEA
503.94
USD
BJ221864-100G
BJ221864-25G
Potassium bromide ≥99% (trace metals basis), FT-IR Grade, Fluka™
Potassium bromide
- Spectrophotometric grade reagents and solvents are made specifically for applications using FT-IR
Formula:
KBr Boiling Pt: 1.383 °C (1.013 hPa) Melting Pt: 734 °C Density: 2.73 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011358 CAS Number: 7758-02-3 EINECS: 231-830-3 |
Specification Test Results
Suitability for IR spectroscopy | complying |
Assay | Min. 99 % |
Metallic impurities (Total) | Max. 10000 ppm |
Vapor Pressure | <0.01 mmHg ( 20 °C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...