Potassium bromide 99,5-100,5, Puriss., meetSpec_BP,Ph.Eur.,USP, Fluka™

Supplier: Honeywell Research Chemicals

02110-100G 02110-6X1KG 02110-500G
BJ02110-100GEA 61.46 USD
BJ02110-100G BJ02110-1KG BJ02110-2.5KG BJ02110-25KG BJ02110-500G
Potassium bromide 99,5-100,5, Puriss., meetSpec_BP,Ph.Eur.,USP, Fluka™
Potassium bromide
Formula: KBr
MW: 119 g/mol
Boiling Pt: 1435
Melting Pt: 730
Density: 2.73 g/cm³ (20 °C)
Storage Temperature: Room Temperature
MDL Number: MFCD00011358
CAS Number: 7758-02-3
EINECS: 231-830-3
ADR: 1
Merck Index: 13,07701

Order Now

Specification Test Results

Assay (calculated to the dried substance) 99.5 - 100.5 %
Loss on drying (105°C, 3h) Max. 0.3 %
Acidity or alkalinity complying
Iron (Fe) Max. 10 ppm
Mg, earth alkali metals (as Ca) Max. 200 ppm
Heavy metals (as Pb) Max. 0.001 %
Bromate (BrO₃) complying
Chloride (Cl) Max. 0.2 %
Iodide (I) complying
Sulfate (SO₄) Max. 0.005 %
Appearance of the solution complying
Residual solvents complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR