Orthophosphoric acid ≥85.0-88.0%, Puriss., meets analytical specification of BP, FCC, NF, Ph. Eur., Fluka™
Supplier: Honeywell Research Chemicals
|
Danger
|
Synonyms:
Phosphoric acid
04107-6X1L
04107-4X2.5L
04107-5L
BJ04107-1LCS
608.1
USD
BJ04107-1L
BJ04107-2.5L
BJ04107-5L
BJ04107-4X5L
Orthophosphoric acid ≥85.0-88.0%, Puriss., meets analytical specification of BP, FCC, NF, Ph. Eur., Fluka™
Orthophosphoric acid
Formula:
H₃PO₄ Boiling Pt: 158 °C (1.013 hPa) Melting Pt: 21 °C Density: 1.71 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011340 CAS Number: 7664-38-2 EINECS: 231-633-2 UN: 1805 ADR: 8,III |
Specification Test Results
Vapor Pressure | 2.2 mmHg ( 20 °C) |
Assay | 85.0 - 88.0 % |
Volatile acids (CH₃COOH) | Max. 0.001 % |
Arsenic (As) | Max. 0.0002 % |
Cadmium (Cd) | Max. 0.0003 % |
Copper (Cu) | Max. 0.001 % |
Iron (Fe) | Max. 0.001 % |
Mercury (Hg) | Max. 0.0001 % |
Lead (Pb) | Max. 0.0003 % |
Zinc (Zn) | Max. 0.001 % |
Heavy metals (as Pb) | Max. 0.0005 % |
Chloride (Cl) | Max. 0.0005 % |
Fluoride (F) | Max. 0.0005 % |
Nitrate (NO₃) | complying |
Phosphite,Hypoph. (H₃PO₃) | Max. 0.003 % |
Phosphite, hypophosphite | complying |
Sulfate (SO₄) | complying |
Appearance of the solution | complying |
Substances precipitated by NH₃ | complying |
Alkali phosphates | complying |
Residual solvents | complying |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...