Nitric acid ≥65% Titrimetric, Puriss. p.a. Hg_0,0000005, Fluka™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Nitric acid solution

84378-1L 84378-2.5L
BJ84378-1LEA 200.35 USD
BJ84378-1L BJ84378-2.5L
Nitric acid ≥65% Titrimetric, Puriss. p.a. Hg_0,0000005, Fluka™
Nitric acid
Formula: HNO₃
MW: 63.01
Boiling Pt: 87
Melting Pt: -42
Density: 1.400…1.480 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011349
CAS Number: 7697-37-2
EINECS: 231-714-2
UN: 2031
ADR: 8 II
Merck Index: 13,06610

Order Now

Specification Test Results

Ignition Residue ≤0.0005% (as SO₄)
Assay 65.0 - 66.0 %
Density (D 20/4) 1.390 - 1.410
Residue on ignition (SO₄) Max. 5 ppm
Silver (Ag) Max. 0.01 ppm
Arsenic (As) Max. 0.01 ppm
Aluminium (Al) Max. 0.05 ppm
Barium (Ba) Max. 0.01 ppm
Bismuth (Bi) Max. 0.1 ppm
Calcium (Ca) Max. 0.5 ppm
Cadmium (Cd) Max. 0.01 ppm
Cobalt (Co) Max. 0.01 ppm
Chromium (Cr) Max. 0.02 ppm
Copper (Cu) Max. 0.01 ppm
Iron (Fe) Max. 0.2 ppm
Mercury (Hg) Max. 0.005 ppm
Potassium (K) Max. 0.05 ppm
Lithium (Li) Max. 0.01 ppm
Magnesium (Mg) Max. 0.1 ppm
Manganese (Mn) Max. 0.01 ppm
Molybdenum (Mo) Max. 0.02 ppm
Sodium (Na) Max. 0.5 ppm
Nickel (Ni) Max. 0.02 ppm
Lead (Pb) Max. 0.01 ppm
Strontium (Sr) Max. 0.01 ppm
Thallium (Tl) Max. 0.05 ppm
Zinc (Zn) Max. 0.05 ppm
Chloride (Cl) Max. 0.5 ppm
Phosphate (PO₄) Max. 0.5 ppm
Sulfate (SO₄) Max. 1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR