Murexide for complexometry, Fluka™

Supplier: Honeywell Research Chemicals

Synonyms: Ammonium purpurate, Murexide triturate, Ammonium 5-(2,4,6-trioxoperhydropyrimidin-5-ylideneamino)barbiturate, Purpuric acid ammonium salt

33414-10G 33414-25G 33414-5G
BJ33414-10GEA 100.67 USD
BJ33414-10G BJ33414-25G BJ33414-5G
Murexide for complexometry, Fluka™
Murexide
Formula: C₈H₈N₆O₆
MW: 284.19 g/mol
Melting Pt: 300
Storage Temperature: Ambient
MDL Number: MFCD00012777
CAS Number: 3051-09-0
EINECS: 221-266-6
Merck Index: 13,06330

Order Now

Specification Test Results

Suitability for Complexometry complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR