Molecular sieve A4 (0.4 nm, 4 Å), pellets, Diameter 3.2 mm, Fluka™

Supplier: Honeywell Research Chemicals

Synonyms: Molecular sieve 0.4 nm, Molecular sieves pack 4A, Linde 4A, Sodium aluminium silicate

334294-1KG 334294-5KG
BJ334294-1KGEA 207 USD
BJ334294-1KG BJ334294-5KG
Molecular sieve A4 (0.4 nm, 4 Å), pellets, Diameter 3.2 mm, Fluka™
Molecular sieve A4 (0.4 nm, 4 Å)
Melting Pt: 400 °C
Density: 2.0 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD03457537
CAS Number: 70955-01-0

Order Now

Specification Test Results

Diameter 3.2 mm
Form pellets
Granulation complying
Pore Size (Ång Ström) 4


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR