Magnesium oxide light, (dried basis) 98.0-100.5% (dried basis), Puriss., meets analytical specification of BP, Ph. Eur., USP, FCC, E530, Fluka™

Supplier: Honeywell Research Chemicals

Synonyms: Magnesia usta

13138-1KG 13138-500G
BJ13138-1KGEA 323.05 USD
BJ13138-1KG BJ13138-500G
Magnesium oxide light, (dried basis) 98.0-100.5% (dried basis), Puriss., meets analytical specification of BP, Ph. Eur., USP, FCC, E530, Fluka™
Magnesium oxide
Formula: MgO
MW: 40.3 g/mol
Boiling Pt: 3600
Melting Pt: 2800
Density: 3.58 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011109
CAS Number: 1309-48-4
EINECS: 215-171-9
Merck Index: 13,05700

Order Now

Specification Test Results

Assay (calculated to ignited substance) 98.0 - 100.5 %
Loss on ignition Max. 5.0 %
In water soluble matter Max. 1.0 %
Insoluble in acetic acid Max. 0.1 %
Insoluble in HCl Max. 0.1 %
Arsenic (As) Max. 2 ppm
Calcium (Ca) Max. 1 %
Copper (Cu) Max. 0.001 %
Iron (Fe) Max. 0.02 %
Lead (Pb) Max. 2 ppm
Zinc (Zn) Max. 0.001 %
Heavy metals (as Pb) Max. 20 ppm
Chloride (Cl) Max. 0.05 %
Sulfate (SO₄) Max. 0.5 %
Free alkali + Soluble salts complying
Appearance of the solution complying
Bulk Density Max. 0.15 g/ml
Residual solvents complying
Filling Volume ≤0.15 g/mL


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR