Lactic acid ≥99%, tested according to Ph. Eur., Fluka™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: DL-Lactic acid, 2-Hydroxypropionic acid, 2-Hydroxypropanoic acid

69775-1L 69775-250ML
BJ69775-1LEA 253.38 USD
BJ69775-1L BJ69775-250ML
Lactic acid ≥99%, tested according to Ph. Eur., Fluka™
Lactic acid
Formula: H₃CCH(OH)COOH
MW: 90.08 g/mol
Boiling Pt: 122 °C (20 hPa)
Density: 1.209 g/cm³
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00004520
CAS Number: 50-21-5
Merck Index: 13,05351

Order Now

Specification Test Results

Appearance of the substance complying
Assay of total acid 88.0 - 92.0 %
Sulfated ash Max. 0.1 %
Calcium (Ca) Max. 200 ppm
Heavy metals (as Pb) Max. 10 ppm
Sulfate (SO₄) Max. 200 ppm
Citronellic-, Oxalic- and Phosphoric acid complying
Sugar and other reducing substances complying
In ether insoluble matter complying
Residual solvents complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR