Iron(II) chloride tetrahydrate ≥99.0% (RT), Puriss. p.a., Fluka™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Ferrous chloride tetrahydrateIron dichloride tetrahydrate

44939-250G 44939-1KG 44939-50G
BJ44939-250GEA 119.83 USD
BJ44939-250G BJ44939-1KG BJ44939-50G
Iron(II) chloride tetrahydrate ≥99.0% (RT), Puriss. p.a., Fluka™
Iron (II) chloride tetrahydrate
Formula: FeCl₂·4H₂O
MW: 198.81 g/mol
Melting Pt: 105…110 °C
Storage Temperature: Ambient
MDL Number: MFCD00149709
CAS Number: 13478-10-9
UN: 3260
ADR: 8,III

Order Now

Specification Test Results

Assay 99.0 - 101.0 %
Arsenic (As) Max. 5 ppm
Calcium (Ca) Max. 10 ppm
Cadmium (Cd) Max. 5 ppm
Cobalt (Co) Max. 50 ppm
Chromium (Cr) Max. 10 ppm
Copper (Cu) Max. 5 ppm
Iron(III) Max. 2000 ppm
Potassium (K) Max. 50 ppm
Magnesium (Mg) Max. 5 ppm
Manganese (Mn) Max. 50 ppm
Sodium (Na) Max. 50 ppm
Nickel (Ni) Max. 50 ppm
Lead (Pb) Max. 10 ppm
Zinc (Zn) Max. 20 ppm
Sulfate (SO₄) Max. 100 ppm
Total nitrogen (as N) Max. 10 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR