Iodine 99,8-100,5, Puriss., meetSpec_BP,Ph.Eur.,USP, Fluka™

Supplier: Honeywell Research Chemicals
Danger

03002-6X1KG 03002-500G
BJ03002-1KGCS 3291.24 USD
BJ03002-1KG BJ03002-500G
Iodine 99,8-100,5, Puriss., meetSpec_BP,Ph.Eur.,USP, Fluka™
Iodine
Formula: I₂
MW: 253.81 g/mol
Boiling Pt: 184.3
Melting Pt: 114
Density: 4.930 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011355
CAS Number: 7553-56-2
EINECS: 231-442-4
UN: 3495
ADR: 8 III
Merck Index: 13,05036

Order Now

Specification Test Results

Vapor Density 9 (vs air)
Assay 99.8 - 100.5 %
Non-volatile matter Max. 0.03 %
Bromide, chloride (as Cl) Max. 0.01 %
Residual solvents complying
Vapor Pressure 0.31 mmHg ( 25 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR