Hydrochloric acid fuming ≥37%, Puriss. p.a. Reag. Ph. Eur., ACS, ISO APHA: ≤10, Fluka™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Hydrochloric acid solution, Muriatic acid

30721-6X1L 30721-6X1L-GL 30721-2.5L 30721-4X2.5L 30721-2.5L-GL
BJ30721-1LCS 472.15 USD
BJ30721-1L BJ30721-1L-GL BJ30721-2.5L BJ30721-4X2.5L BJ307212.5LGL
Hydrochloric acid fuming ≥37%, Puriss. p.a. Reag. Ph. Eur., ACS, ISO APHA: ≤10, Fluka™
Hydrochloric acid
Formula: HCl
Boiling Pt: 42 °C (1.013 hPa)
Melting Pt: –35 °C
Density: 1.190 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II

Order Now

Specification Test Results

Assay 37 - 38.0 %
Density (20°C) Min. 1.18 g/ml
Residue on ignition (SO₄) Max. 5 ppm
Non-volatile matter Max. 0.005 %
Ammonium (NH₄) Max. 1 ppm
Silver (Ag) Max. 0.01 ppm
Aluminium (Al) Max. 0.000005 %
Arsenic (As) Max. 0.01 ppm
Barium (Ba) Max. 0.000002 %
Beryllium (Be) Max. 0.000002 %
Bismuth (Bi) Max. 0.1 ppm
Calcium (Ca) Max. 0.00005 %
Cadmium (Cd) Max. 0.01 ppm
Cobalt (Co) Max. 0.000001 %
Chromium (Cr) Max. 0.000002 %
Copper (Cu) Max. 0.02 ppm
Iron (Fe) Max. 0.2 ppm
Germanium (Ge) Max. 0.000005 %
Mercury (Hg) Max. 0.01 ppm
Potassium (K) Max. 0.00001 %
Lithium (Li) Max. 0.000001 %
Magnesium (Mg) Max. 0.00001 %
Manganese (Mn) Max. 0.000001 %
Molybdenum (Mo) Max. 0.02 ppm
Sodium (Na) Max. 0.00005 %
Nickel (Ni) Max. 0.000002 %
Lead (Pb) Max. 0.02 ppm
Strontium (Sr) Max. 0.000001 %
Titanium (Ti) Max. 0.00001 %
Thallium (Tl) Max. 0.000005 %
Vanadium (V) Max. 0.000001 %
Zinc (Zn) Max. 0.000005 %
Zirconium (Zr) Max. 0.00001 %
Heavy metals (ICP) ACS Max. 1 ppm
Free chlorine (Cl) Max. 0.5 ppm
Bromide (Br) Max. 0.005 %
Phosphate (PO₄) Max. 0.00005 %
Sulfate (SO₄) Max. 1 ppm
Sulfite (SO₃) Max. 1 ppm
Appearance of the substance complying
APHA Max. 10
Residual solvents complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR