Hydrochloric acid 37% ACS, Fluka™
Supplier: Honeywell Research Chemicals
|
Danger
|
Synonyms:
Hydrochloric acid solution, Muriatic acid
258148-2.5L
258148-4L
258148-4X4L
258148-6X500ML
BJ258148-2.5LEA
289
USD
BJ258148-2.5L
BJ258148-4L
BJ258148-4X4L
BJ258148-500ML
Hydrochloric acid 37% ACS, Fluka™
Hydrochloric acid
Formula:
HCl MW: 36.46 Boiling Pt: 42 °C (1.013 hPa) Melting Pt: –35 °C Density: 1.190 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011324 CAS Number: 7647-01-0 EINECS: 231-595-7 UN: 1789 ADR: 8 II |
Specification Test Results
Assay | 36.5 - 38.0 % |
Residue on ignition (SO₄) | Max. 5 ppm |
Arsenic (As) | Max. 0.01 ppm |
Iron (Fe) | Max. 0.2 ppm |
Vapor Pressure | 3.23 psi ( 21.1 °C) |
Vapor Density | 1.3 (vs air) |
Heavy metals (ICP) ACS | Max. 1 ppm |
Ammonium (NH₄) | Max. 1 ppm |
Bromide (Br) | Max. 50 ppm |
Free chlorine (Cl) | Max. 1 ppm |
Sulfate (SO₄) | Max. 1 ppm |
Sulfite (SO₃) | Max. 1 ppm |
APHA | Max. 10 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...