Hydrobromic acid 48% ACS, Fluka™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Hydrogen bromide

244260-1L 244260-500ML
BJ244260-100MLEA 89.67 USD
BJ244260-100ML BJ244260-1L BJ244260-2.5L BJ244260-500ML
Hydrobromic acid 48% ACS, Fluka™
Hydrogen bromide
Formula: HBr
Boiling Pt: 126 °C (1.013 hPa)
Melting Pt: –11 °C
Density: 1.490 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011323
CAS Number: 10035-10-6
UN: 1788
ADR: 8,II

Order Now

Specification Test Results

Vapor Pressure 320 psi ( 21.1 °C)
Assay 47.0 - 49.0 %
Residue on ignition (SO₄) Max. 0.002 %
Iron (Fe) Max. 1 ppm
Ignition Residue ≤0.002%
Selenium (Se) Max. 0.01 ppm
Heavy metals (ICP) ACS Max. 5 ppm
Chloride (Cl) Max. 0.05 %
Iodide (I) Max. 0.003 %
Phosphate (PO₄) Max. 0.001 %
Sulfate and sulfite (as SO₄) Max. 0.003 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR