Glycerine, anhydrous 99.0-101.0% (by alkalimetry), Puriss., meets analytical specification of BP, FCC, Ph. Eur., USP, E422, Riedel-de Haën™

Supplier: Honeywell Research Chemicals

Synonyms: Glyceritol, Glysanin, 1,2,3-Propanetriol, Osmoglyn, Glycerin, Glycyl alcohol, Glyrol, Propane-1,2,3-triol, Propanetriol, Trihydroxypropane, Glycerol

15523-4X5L 15523-250KG 15523-6X1L
BJ15523-1LCS 782.38 USD
BJ15523-1L BJ15523-5L BJ15523-250KG BJ15523-6X1L
Glycerine, anhydrous 99.0-101.0% (by alkalimetry), Puriss., meets analytical specification of BP, FCC, Ph. Eur., USP, E422, Riedel-de Haën™
Glycerine

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.


Beilstein Number: 635685

Formula: HOCH₂CH(OH)CH₂OH
MW: 92.09 g/mol
Boiling Pt: 290
Melting Pt: 18.6
Density: 1.26 g/cm³ (20 °C)
Flash Pt: 160 °C
Storage Temperature: Ambient
MDL Number: MFCD00004722
CAS Number: 56-81-5
EINECS: 200-289-5
Merck Index: 13,04497
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3668cce85af1464db21fd098526252076b567956&vid=86a15453b54e7106c121d32e4e716385003c4507&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity (IR) complying
Assay (alkalimetric) 99.0 - 101.0 %
Assay (calculated on the anhydrous basis) 99.0 - 101.0 %
Density (D 20/20) 1.2580 - 1.2644
Density (D 25/25) Min. 1.259
Refractive index (n 20/D) 1.471 - 1.475
Water (Karl Fischer) Max. 1.0 %
Sulfated ash Max. 0.01 %
Free acid (as CH₃COOH) Max. 0.003 %
Acid. or alk. react. impurities complying
Arsenic (As) Max. 1 ppm
Cadmium (Cd) Max. 1 ppm
Copper (Cu) Max. 0.001 %
Mercury (Hg) Max. 1 ppm
Lead (Pb) Max. 1 ppm
Zinc (Zn) Max. 0.001 %
Heavy metals (as Pb) Max. 5 ppm
Chloride (Cl) Max. 10 ppm
Sulfate (SO₄) Max. 10 ppm
Halogene compounds (as Cl) Max. 30 ppm
Reaction against H₂SO₄ complying
Aldehydes (as HCHO) Max. 10 ppm
Acrolein, Glucose + NH4-compounds complying
Ester complying
Fatty acids and ester complying
Sugar complying
Impurity A + related substances (GC) complying
Other impurities (GC) total Max. 1.0 %
Other Single impurities (GC) Max. 0.1 %
1,2,4-butantriole (GC) Max. 0.2 %
Diethylene glycole (GC) Max. 0.10 %
Ethylene glycole (GC) Max. 0.10 %
3-monochloropropane-1.2-diole Max. 0.1 ppm
Residual solvents complying
Colour complying
Appearance of the solution complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR