Disodium hydrogen phosphate heptahydrate 98.0-102.0% ACS, Fluka™

Supplier: Honeywell Research Chemicals

Synonyms: Sodium phosphate dibasic heptahydrateSodium monohydrogen phosphate heptahydrateDisodium hydrogenorthophosphate 7-hydrateDisodium hydrogenorthophosphate heptahydrateSodium phosphate dibasic 7-hydrateDisodium phosphate (heptahydrate)di-Sodium hydrogen phosphate 7-hydrateSodium monohydrogen phosphate 7-hydrateSodium hydrogen phosphate heptahydrate

S9390-1KG S9390-6X500G
BJS9390-1KGEA 188.79 USD
BJS9390-1KG BJS9390-500G
Disodium hydrogen phosphate heptahydrate 98.0-102.0% ACS, Fluka™
di-Sodium hydrogen phosphate heptahydrate
Formula: HNa₂O₄P·7H₂O
MW: 268.07 g/mol
Boiling Pt: Decomposes on heating
Storage Temperature: Ambient
MDL Number: MFCD00149180
CAS Number: 7782-85-6
ADR: 3,

Order Now

Specification Test Results

Assay 98.0 - 102.0 %
Water insoluble matter Max. 0.005 %
pH (5 %, 25 °C) 8.7 - 9.3
Iron (Fe) Max. 0.001 %
Heavy metals (as Pb) Max. 0.001 %
Chloride (Cl) Max. 0.001 %
Sulfate (SO₄) Max. 0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR