Disodium hydrogen phosphate dihydrate 98,5-101,0 Dried, Puriss. p.a. Reag. Ph. Eur., Fluka™
Supplier: Honeywell Research Chemicals
Synonyms:
sec-Sodium phosphate, Sodium monohydrogen phosphate dihydrate, Sodium hydrogenphosphate dihydrate, di-Sodium hydrogen orthophosphate dihydrate, Sodium phosphate dibasic dihydrate, Disodium phosphate, di-Sodium hydrogen phosphate dihydrate, Sorensen's salt
30435-6X1KG
30435-500G
30435-5KG
BJ30435-1KGCS
591.1
USD
BJ30435-1KG
BJ30435-500G
BJ30435-5KG
Disodium hydrogen phosphate dihydrate 98,5-101,0 Dried, Puriss. p.a. Reag. Ph. Eur., Fluka™
di-Sodium hydrogen phosphate dihydrate
Formula:
Na₂HPO₄·2H₂O MW: 177.99 g/mol Melting Pt: 92.5 Density: 2.07 g/cm³ (16 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149182 CAS Number: 10028-24-7 |
Specification Test Results
Assay (calculated to the dried substance) | 98.5 - 101.0 % |
Assay of NaH₂PO₄ (calculated to dried substances) | Max. 0.5 % |
Loss on drying (130°C) | 19.5 - 21.0 % |
pH (5 %, 20°C) | 9.0 - 9.2 |
Arsenic (As) | Max. 0.00005 % |
Iron (Fe) | Max. 0.0005 % |
Potassium (K) | Max. 0.005 % |
Heavy metals (as Pb) | Max. 0.0005 % |
Chloride (Cl) | Max. 0.001 % |
Sulfate (SO₄) | Max. 0.005 % |
Total N | Max. 0.001 % |
Appearance of the solution | complying |
Reducing impurities | complying |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...