di-Potassium hydrogen phosphate ≥98% ACS, Fluka™
Supplier: Honeywell Research Chemicals
P3786-100G
P3786-1KG
P3786-2.5KG
P3786-500G
P3786-6X500G
BJP3786-100GEA
72.68
USD
BJP3786-100G
BJP3786-1KG
BJP3786-2.5KG
BJP3786-500G
BJP3786-6X500G
di-Potassium hydrogen phosphate ≥98% ACS, Fluka™
di-Potassium hydrogen phosphate
Formula:
K₂HPO₄ Melting Pt: 340 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00011383 CAS Number: 7758-11-4 EINECS: 231-834-5 |
|
Specification Test Results
Assay | Min. 98.0 % |
Water insoluble matter | Max. 0.01 % |
Loss on drying (105°C) | Max. 1.0 % |
pH (5 %, 25 °C) | 8.5 - 9.6 |
Iron (Fe) | Max. 10 ppm |
Sodium (Na) | Max. 500 ppm |
Heavy metals (as Pb) | Max. 5 ppm |
Chloride (Cl) | Max. 30 ppm |
Total N | Max. 10 ppm |
Sulfate (SO₄) | Max. 50 ppm |
Appearance of the solution | complying |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...