Copper(II) oxide ≥99.0% (RT), powder, Puriss. p.a., Fluka™

Supplier: Honeywell Research Chemicals

Synonyms: Copper monoxide granulatedCopper oxideCupric oxide

61200-500G 61200-100G
BJ61200-500GEA 463.97 USD
BJ61200-500G BJ61200-100G
Copper(II) oxide ≥99.0% (RT), powder, Puriss. p.a., Fluka™
Copper (II) oxide
Formula: CuO
MW: 79.55 g/mol
Melting Pt: 1.336 °C
Density: 6.450 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010979
CAS Number: 1317-38-0
EINECS: 215-269-1

Order Now

Specification Test Results

Assay 99.0 - 101.0 %
Insoluble in HCl Max. 200 ppm
Calcium (Ca) Max. 100 ppm
Cadmium (Cd) Max. 10 ppm
Cobalt (Co) Max. 10 ppm
Chromium (Cr) Max. 50 ppm
Iron (Fe) Max. 500 ppm
Potassium (K) Max. 50 ppm
Magnesium (Mg) Max. 100 ppm
Manganese (Mn) Max. 10 ppm
Sodium (Na) Max. 500 ppm
Nickel (Ni) Max. 50 ppm
Lead (Pb) Max. 500 ppm
Zinc (Zn) Max. 100 ppm
Chloride (Cl) Max. 50 ppm
Total S (as SO₄) Max. 500 ppm
Total N Max. 20 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR