Citric acid monohydrate ≥99.9%, tested according to Ph. Eur., Fluka™
Supplier: Honeywell Research Chemicals
|
Warning
|
Synonyms:
2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid, 2-Hydroxy-1,2,3-propanetricarboxilic acid
27491-1KG
27491-25KG
BJ27491-1KGEA
194.46
USD
BJ27491-1KG
BJ27491-25KG
Citric acid monohydrate ≥99.9%, tested according to Ph. Eur., Fluka™
Citric acid monohydrate
Formula:
C₆H₈O₇.H₂O MW: 210.14 g/mol Melting Pt: 135…152 °C Density: 1.54 g/cm³ (20 °C) Flash Pt: 173.9 Storage Temperature: Ambient |
MDL Number:
MFCD00149972 CAS Number: 5949-29-1 Merck Index: 13,02350 |
|
Specification Test Results
Identity (IR) | complying |
Assay (calculated on the anhydrous basis) | 99.5 - 100.5 % |
Appearance of the solution | complying |
Sulfated ash | Max. 0.1 % |
Water (Karl Fischer) | 7.5 - 9.0 % |
Heavy metals (as Pb) | Max. 10 ppm |
Sulfate (SO₄) | Max. 150 ppm |
Reaction against H₂SO₄ | complying |
Readily carbonisable substances | complying |
Oxalate (as Oxalic acid) | Max. 360 ppm |
Residual solvents | complying |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...