Calcium chloride, anhydrous ≥97%, powder, Fluka™

Supplier: Honeywell Research Chemicals
Warning

Synonyms: Chloro calcium, Calcium dichloride, CaCl2

C4901-1KG C4901-100G C4901-2.5KG C4901-500G
BJC4901-1KGEA 322.3 USD
BJC4901-1KG BJC4901-100G BJC4901-2.5KG BJC4901-500G BJC4901-5KG
Calcium chloride, anhydrous ≥97%, powder, Fluka™
Calcium chloride
Formula: CaCl₂
MW: 110.98 g/mol
Boiling Pt: 1600
Melting Pt: 772
Density: 2.150 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010903
CAS Number: 10043-52-4
EINECS: 233-140-8
Merck Index: 14,01659

Order Now

Specification Test Results

Colour of the solution complying
Clearness of the solution complying
Assay Min. 97.0 %
Mg and alkali salts Max. 3.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR