Bromothymol blue sultone form ACS, Reag. Ph. Eur. indicator, Fluka™

Supplier: Honeywell Research Chemicals

Synonyms: 3,3-Bis(3-bromo-4-hydroxy-5-isopropyl-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide3′,3″-Dibromothymolsulfonphthalein3′,3″-Dibromothymolsulphonphthalein

32714-5G 32714-25G 32714-1KG
BJ32714-1KGEA 2467.61 USD
BJ32714-1KG BJ32714-25G BJ32714-5G
Bromothymol blue sultone form ACS, Reag. Ph. Eur. indicator, Fluka™
Bromothymol blue
Formula: C₂₇H₂₈Br₂O₅S
MW: 624.39 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00005872
CAS Number: 76-59-5
EINECS: 200-971-2

Order Now

Specification Test Results

Suitability as indicator complying
Test of sensitivity complying
Clearness of the solution complying
Change Range (pH6,0-7,4) complying
Curve course (UV) complying

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR